Learn More
Thermo Scientific™ Abgene™ 48 Well 6mL Polypropylene Storage Plate
The 6 mL 48-well storage plate is ideal for storage when large sample volumes are required.
Brand: Thermo Scientific™ AB-0988
316.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 03
Additional Details : Weight : 6.34500kg
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Abgene 48 Well 6mL Polypropylene Storage Plate are manufactured in a Class 100,000 cleanroom environment to ensure that each plate is contamination-free.
Highlights
- 6 mL working volume when sealed with adhesive or heat seals
- 48 square wells
- Pyramidal well bottom for ease of sample retrieval
- Support and stacking ribs
- Angled well corners reduce the capillary action of liquids, preventing cross-contamination between wells
- Sample storage
- Cell culture
- Assay setup; diluting and aliquoting
Compatible Sealing Film:Adhesive and Heat
Hazardous: No
Max Centrifugal Force: Not suitable for centrifugation
Max Vol (Cap Seal): n/a
Max Vol (Film Seal): 6 mL
Max Vol (Mat Seal): n/a
Storage Condition: Ambient
Specifications
Custom Barcoding Available | |
48-well | |
6 x 8 | |
Individually Wrapped | |
Abgene | |
48-well plate | |
Polypropylene | |
48 | |
Cleanroom | |
Square, Conical Bottom (V-well) | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
1 | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
48-well Polypropylene Sample Processing & Storage Plates | |
Autoclavable | |
Low binding PP | |
Natural | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
6000 μL | |
5500 μL | |
Non-treated | |
Yes | |
50 | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |