Learn More
Thermo Scientific™ Polystyrene 96-Well Plate Cover with Notches
Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking
Brand: Thermo Scientific™ AB-0751
66.34 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 03
Additional Details : Weight : 0.88200kg
Description
Polystyrene or polypropylene 96-well plate covers, with or without ridges for easy stacking
Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates. The covers are made of polystyrene or polypropylene and are either ridged or non-ridged.
Highlights
- Polystyrene covers can be used with all 96-well plates
- Ridged covers for stacking of covered plates
- Non-ridged covers for use with automated systems
- Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates
Hazardous: No
Storage Condition: Ambient
Specifications
Clear | |
5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm) | |
Cleanroom | |
Without stacking ridge (for automated systems) | |
50 | |
5 x 3.31 in. (126 x 84 mm) | |
Microplate Lid, cover | |
None | |
All well shapes | |
H1 Corner |
Microplate Lid | |
Polystyrene | |
96 | |
No | |
2 | |
Lid | |
Abgene | |
-20°C to +120°C | |
Abgene 96 and 384-well Plate | |
PCR Ready |