missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene™ Plate Covers
GREENER_CHOICE

Polypropylene 96-well plate covers, with or without ridges for easy stacking

Brand:  Thermo Scientific™ AB-0755

79.69 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


Product Code. 11560304

  • £228.00 / Pack of 50
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Polypropylene 96-well plate covers, with or without ridges for easy stacking

Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates.

Highlights

  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates

Hazardous: No

Storage Condition: Ambient

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm)
Cleanroom
With stacking ridge
50
5 x 3.31 in. (126 x 84 mm)
Microplate Lid, cover
None
All well shapes
H1 Corner
Microplate Lid
Polypropylene
96
No
2
Lid
Abgene
-20°C to +120°C
Abgene 96 and 384-well Plate
PCR Ready
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Abgene™ Plate Covers > For 384-well Thermo-Fast plates; Polypropylene; With stacking ridge

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.