missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Abgene Storage Plate Caps, Strips of 8
GREENER_CHOICE

Storage Plate Caps and Mats are available in a variety of formats.

Brand:  Thermo Scientific™ AB-0981

33.90 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10476113

  • £77.45 / Pack of 120
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Storage Plate Caps and Mats are available in a variety of formats.

Thermo Scientific storage plate cap strips are suitable for use on with Round Well Storage Plates and feature a low-profile design. They come in an 8-cap strip and can simultaneously seal up to 8 well. Each cap is individually numbered.

Highlights

  • 8-cap strip
  • Each cap is individually numbered
  • Simultaneous sealing of 8 wells
  • End tabs for ease of removal
  • Low profile for increased well volume

Autoclavable- Resistance: Not Recommended
DMSO (100%) - Resistance: Successfully Tested
Ethanol (100%) - Resistance: Successfully Tested
Gamma Irradiation - Resistance: Successfully Tested
Hazardous: No
Isopropanol (100%) - Resistance: Successfully Tested
Long Term Strorage: Successfully Tested
Peelable: Successfully Tested
Pierceable:Not Recommended
Resealable: Successfully Tested
Sealing Temp Range: -20°C to +55°C
Storage Condition: Ambient
UV Irradiation- Resistance: Successfully Tested

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Natural
Cleanroom
8
Abgene
No
Abgene 96 Round Well Plate
Polypropylene
Autoclavable
Plate Lid
Yes
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers