missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperPlate PCR Plates, 96-well

96-well PCR plates with a flat deck for improved sealing in PCR and qPCR applications.

£113.00 - £134.33

Specifications

Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Material Polypropylene
No. of Wells 96
View More Specs

Products 6
Product Code Brand Color Skirt Style Product Type Barcode Price Quantity & Availability  
Product Code Brand Color Skirt Style Product Type Barcode Price Quantity & Availability  
10124183
View Documents
Thermo Scientific™
AB-2100
Clear No Barcode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10183653
View Documents
Thermo Scientific™
AB-2100/W
White No Barcode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10657513
View Documents
Thermo Scientific™
AB-2800/B
Blue Full Raised Deck No Barcode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11802133
View Documents
Thermo Scientific™
AB2800Y
Yellow Full Raised Deck No Barcode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10681331
View Documents
Thermo Scientific™
BC-2800
Clear This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10241310
View Documents
Thermo Scientific™
BC2100
Clear Semi Raised Deck Includes Barcode This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The Thermo Scientific 96-Well Semi-Skirted Flat Deck PCR Plates are compatible with most thermal cyclers and ABI platforms, including sequencers, with no adapters necessary. The Thermo Scientific PCR plastics are designed, manufactured, and tested to ensure PCR performance. The plates have 0.3 mL maximum well volume when used with adhesive and heat seals.
The flat plate deck design ensures easy handling, and the plates feature a raised rim to prevent evaporation. The 96-Well Semi-Skirted Flat Deck PCR Plates have a cut corner at the A12 location and have thin well walls for consistent heat transfer.

Highlights
  • Directly compatible with all major major thermal cyclers, including ABI platforms and sequencers with no adapters necessary
  • Flat plate deck facilitates sealing and handling
  • Available as SuperPlate providing 4x more rigidity for superior robotic handling
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Black lettered versions feature black alpha-numeric lettering to allow for easy well identification
  • White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
  • Cut corner: A12
Applications
  • PCR
  • qPCR/Real-Time PCR
  • Sequencing

Hazardous: No
Storage Condition: Ambient

Specifications

Specifications

DNA, DNase, RNase-free
8 x 12
96
Yes
Standard
Polypropylene
For improved sealing in PCR and qPCR applications
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.