Learn More
Thermo Scientific™ Barnstead™ Easypure™ II Compact Ultrapure Water System Filters and Cartridges
£132.52 - £174.68
Product Code | Brand | For Use With (Equipment) | Type | Resin Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Resin Type | Price | Quantity & Availability | |||
10032283
|
Thermo Scientific™
D50229 |
Easypure II Water Purification System, Organic Free/Organic Removal | Cartridge and Filter | Ultrapure Mixed Bed |
£174.68
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
10669532
|
Thermo Scientific™
D50231 |
Easypure II Compact Ultrapure Water System, Organic/Ion Exchange Cartridge | Cartridge and Filter | Activated Carbon and Mixed Bed Ion Exchange Resin |
£132.52
Each |
In Stock
Log in to see stock. |
|||
Description
- Easypure™ II Systems require pretreated feedwater
- From the chart, select appropriate cartridges based on application and feedwater type
Order Info
Cartridges and filters must be ordered separately
Specifications
Cartridge and Filter |