Learn More
Thermo Scientific™ BetaSil™ C18 Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 70105-012106
 View more versions of this product
Product Code. 11548720
  • £428.13 / Pack of 4
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Betasil C18 Solid Phase
  • Ligand on 100Å silica
  • High surface area with high bonded phase coverage
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
2.1 mm
BetaSil
10 mm
C18