missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ AX Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
£315.18 - £458.70
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
11533301
|
Thermo Scientific™
73105-022106 |
2.1 mm | 20 mm |
£458.70
Pack of 4 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||||
11523301
|
Thermo Scientific™
73105-014006 |
4 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11513301
|
Thermo Scientific™
73105-012106 |
2.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Weak anion exchange phase for multiple charged species
- 300Å pore size for enhanced protein and peptide separations
- Suitable for HILIC retention and separation of highly polar molecules
- Superb stability under demanding pH conditions
Specifications
5 μm | |
AX |