Learn More
Unity Lab Services™ Thermo Scientific™ Biological Safety Cabinets and Clean Benches Validation Documention for Thermo Scientific™ HeraGuard Eco
Product Code. 15530230
Product Code. 15530230
missing translation for 'mfr': Unity Lab Services™ IOQDOCE89003486
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Biological Safety Cabinets and Clean Benches Validation Documention for Thermo Scientific HeraGuard Eco
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Biological Safety Cabinets and Clean Benches including Thermo Scientific HeraGuard Eco
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Filter leakage, Air Velocity, Electrical Safety, Air-Flow Smoke Pattern, KI-DISCUS testing.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation