Learn More
Unity Lab Services™ Thermo Scientific Centrifugation Validation Documention for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
OEM service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Centrifugation Validation Documention for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
Brand: Unity Lab Services™ IOQDOCE89003492
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Thermo Scientific Centrifugation including Benchtop, Micro, Bioprocessing, Clinical, and Standard floormodel centrifuges: Thermo Scientific, Sorvall | |
Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models). | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
IQ/OQ Documentation |