Learn More
Unity Lab Services™ Thermo Scientific™ Centrifugation Validation Service
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
Brand:  Unity Lab Services™ IOQPCKE89003492
1632.85 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Product Code. 15536209
  • £1921.00 / Each
Explore more special offers
Description
Description
IQ
  • Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
  • System Component Information Detailed configuration information for each system component have been recorded.
  • OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
  • Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
Key Tests: Min/Max Speed, Brief/Extended Time, Temperature for Refrigerated Models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermo Scientific Centrifugation including Benchtop, Micro, Bioprocessing, Clinical, and Standard floormodel centrifuges: Thermo Scientific, Sorvall
Industry-Exclusive, No-Charge Requalification Guarantee
Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service
Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models).
Contact your local sales representative for a list of standard and extended coverage areas.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products