missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Clear Seal Heat Sealing Film
Ideal for applications requiring pierceable non-conductive seal material
Brand: Thermo Scientific™ AB-3797
1209.73 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for applications requiring pierceable non-conductive seal material
- Suitable for PCR applications with screw- or clip-down heated lid thermal cyclers only
- Seals polypropylene, polystyrene, polyethylene and Diamond plates
- Easily pierced and peelable by hand from 96-well plates
- Temperature range: -80°C to +80°C

Specifications
Clear | |
Clear Seal Thin, Polymer, 78mm x 610m | |
Non-sterile | |
96 | |
1 Roll | |
Storage, PCR, qPCR, Automation, ELISA, Molecular Diagnostics | |
ALPS 5000 & ALPS 3000 | |
Abgene | |
No | |
-80°C to +80°C, -80°C to +120°C (in Cycler) | |
Polypropylene, Polyethylene, Polystyrene |
Life Science Reagent Resistant | |
Polyethylene | |
Not Autoclavable | |
Clear Seal Thin, Polymer, Roll 78mm x 610m | |
610 m x 78 mm | |
Roll | |
Heat Seal | |
Polypropylene | |
Yes | |
All well shapes |
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Clear Seal Heat Sealing Film > For ALPS 300 and 3000; Roll
Spot an opportunity for improvement?Share a Content Correction