Learn More
Unity Lab Services™ Thermo Scientific™ CO2 Incubators Validation Documention for Thermo Scientific™ CO2 Incubators
Product Code. 15145873
Product Code. 15145873
Brand: Unity Lab Services™ IOQDOCE89003500
Edge
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific CO2 Incubators Validation Documention for Thermo Scientific CO2 Incubators
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ThermoScientific ULT Freezers including TDE series, FDE series, RDE series, IUE series
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation