missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Easy Pierce Heat Sealing Foil
Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
£171.00 - £176.47
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11918935
|
Thermo Scientific™
AB-1720 |
EasyPierce 20μm Thin Foil; 85x135mm |
£176.47
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11979355
|
Thermo Scientific™
AB-0757 |
Easy Pierce Foil, 85 x 135mm |
£171.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
11540284
|
Thermo Scientific™
AB-0745 |
Easy Peel film laminate, 85 x 135 mm |
£176.47
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.
- Suitable for PCR (requires the pressure from a screw-down heated lid)
- Easily pierced
- Peelable bond can be removed by hand
- 20μm foil is even more easily pierced and is suitable for multiple layering
- Recommended for use with DMSO
- Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)