Learn More
Thermo Scientific™ Lysis Buffer for MagJET Blood RNA Kit
Generate sample lysate in order to isolate total RNA. Thermo Scientific™ MagJET™ Lysate Buffer is a component of the MagJET™ Whole Blood RNA Kit (K2751) and may be purchased separately when additional solution is needed. It is provided in 27mL bottles.
Brand: Thermo Scientific™ R2391
54.86 GBP valid until 2024-12-31
Use promo code "21969" to get your promotional price.
Code : 88
Additional Details : Weight : 0.07000kg
Description
The Thermo Scientific™ MagJET™ Whole Blood RNA Kit is designed for fast and efficient purification of total RNA from fresh or frozen whole blood treated with EDTA or citrate, blood fractions, bone marrow, or samples stabilized with PAXgene™ or Tempus™. The kit utilizes paramagnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality RNA is free of proteins, nucleases and other contaminants or inhibitors.
Specifications
Lysis Buffer | |
MagJET | |
27 mL |
Safety and Handling
- Lysis Buffer for MagJET Blood RNA Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Thiocyanic acid
For Research Use Only. Not for use in diagnostic procedures.