Learn More
Thermo Scientific™ dsDNase
dsDNase is a heat inactivatable recombinant shrimp DNase used to remove genomic DNA contamination of RNA samples before reverse transcription.
Brand: Thermo Scientific™ EN0771
123.38 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific dsDNase is an engineered shrimp DNase designed for rapid and safe removal of contaminating genomic DNA from RNA samples. It is an endonuclease that cleaves phosphodiester bonds in DNA to yield oligonucleotides with 5'-phosphate and 3'-hydroxyl termini. Highly specific activity towards double-stranded DNA ensures that RNA and single-stranded DNA such as cDNA and primers are not cleaved. dsDNase is easily inactivated by modetate heat treatment (55°C). These features make dsDNase an excellent choice for gDNA elimation prior reverse transcription. It allows for dramatically simplified workflow which combines genomic DNA elimination and cDNA synthesis into one-tube procedure.
Highlights
- Highly specific degradation of double-stranded DNA
- Fast 2 min protocol for gDNA removal prior reverse transcription
- Heat-labile - irreversibly inactivated by moderate heat treatment (55°C)
- Genomic DNA removal from RNA samples prior first strand cDNA synthesis, RT-PCR and RT-qP
Specifications
DNase |
For Research Use Only. Not for use in diagnostic procedures.