missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET Plasmid DNA Kit

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

£167.00 - £554.00

Specifications

Format Kit
Isolation Technology Magnetic Bead
Sample Type Bacteria
For Use With (Application) Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
High-throughput Compatibility High-throughput Compatible, Not High-throughput Compatible (Manual)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13565180
View Documents
Thermo Scientific™
K2791
96 Preps
£167.00
96 tests
Please to purchase this item. Need a web account? Register with us today!
13585180
View Documents
Thermo Scientific™
K2792
4 x 96 Preps
£554.00
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific MagJET Plasmid DNA Kit, sufficient for 96 preps, is designed for fast and efficient purification of plasmid DNA (up to 4–5 μg/mL) or high copy plasmid DNA from overnight E. coli culture.

The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET Magnetic Beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification for low sample throughput users.

The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors. Purified plasmid DNA can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.

Highlights
High yields—up to 5 μg of plasmid DNA from 1 mL of bacterial culture
Pure—isolated DNA is immediately ready-to-use in downstream application such as sequencing, transcription, digestion with restriction enzymes or transfection of robust cell lines
Flexible—automation and manual processing protocols available
Repeatable results and robust performance

Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
• In vitro transcription
• Transformation
• Transfection of robust cell lines

Specifications

Specifications

Kit
Bacteria
High-throughput Compatible, Not High-throughput Compatible (Manual)
MagJET
Plasmid DNA
Magnetic Bead
Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR
< 100 μg (Small-Scale) Plasmid DNA
Plasmid DNA Kit
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.