missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MagJET™ Whole Blood gDNA Kit

Purifies genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, using magnetic bead technology.

£275.00

Specifications

Content And Storage Store Proteinase K solution at room temperature as long as the vial remains sealed. After opening, store at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
For Use With (Application) PCR and qPCR, FastDigest and conventional restriction digestion, Southern Blotting
View More Specs

Products 1
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
Product Code Brand Includes No. of Reactions Price Quantity & Availability  
13595170
View Documents
Thermo Scientific™
K2741
Lysis Buffer (15mL), MagJET Magnetic Beads (2 x 1.4mL), Proteinase K (2 x 1mL), Binding Buffer (23mL), conc. Wash Buffer 1 (100mL), conc. Wash Buffer 2 (40mL), Elution Buffer (30mL) 96 Preps This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

The Thermo Scientific MagJET Whole Blood Genomic DNA Kit is designed for fast and efficient purification of genomic DNA from fresh or frozen whole blood treated with EDTA or citrate, as well as blood fractions. Note: Samples preserved with heparin are not compatible with this kit.
The kit utilizes paramagnetic bead technology enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users.
The resulting high quality DNA is free of proteins, nucleases and other contaminants or inhibitors, can be used in a wide range of downstream applications such as PCR, qPCR and other enzymatic reactions.

Highlights
  • Compatible - suitable to use with common stabilizers such as EDTA, citrates
  • Versatile - suitable for gDNA purification from fresh, frozen or dried blood, as well as bone marrow and buffy coat
  • Efficient - high yields of pure DNA free of contaminants and downstream application inhibitors
  • Flexible - easily adjustable for high throughput automation platforms and manual processing
Applications
Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
  • PCR and qPCR
  • FastDigest and conventional restriction digestion
  • Southern blotting
Includes
  • Lysis Buffer
  • MagJET Magnetic Beads
  • Proteinase K
  • Binding Buffer
  • Wash Buffer 1 (conc.)
  • Wash Buffer 2 (conc.)
  • Elution Buffer

Hazardous: Yes
Hazardous: No
Quality Control: The MagJET Whole Blood Genomic DNA Kit is qualified by isolating genomic DNA from 200 μL of frozen human whole blood following the protocol outlined in the manual. The quality of isolated DNA is evaluated spectrophotometrically and by agarose gel electrophoresis. The purified DNA has an A260/A280 ratio of 1.8±0.2. The functional quality of purified DNA is evaluated by digestion with restriction endonucleases.
Shelf Life: 36 Months
Shipping Condition: Ambient
Storage Condition: Ambient

Specifications

Specifications

Store Proteinase K solution at room temperature as long as the vial remains sealed. After opening, store at -20deg.C. MagJET Magnetic Beads should be stored at 4deg.C. Other components of the kit should be stored at room temperature (15deg. to 25deg.C).
PCR and qPCR, FastDigest and conventional restriction digestion, Southern Blotting
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 3 Years

SDS
Documents

Documents

Product Certifications