Learn More
Thermo Scientific™ TopVision Agarose Tablets
High quality, multipurpose agarose tablets in blister packaging.
Brand: Thermo Scientific™ R2801
116.72 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.20000kg
Description
Description
TopVision Agarose Tablets, low EEO (LE), is multipurpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.
Features
• Convenient tablet format - no messy weighing required.
• Fast dissolving protocol.
•“Green”- manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
• Enhanced resolution and reduced background.
Applications
Ideal for routine DNA and RNA gel electrophoresis and blotting assays.
Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.
Appearance:White to off-white powder
> Ash:≤ 0.5%
> CAS:9012-36-6
> EEO:≤ 0.13
> Gel Strength:≥ 1200 g/cm2 (1% Gel)
> Gelling Point:36°C ± 1.5°C (1.5% gel)
> Hazardous:No
> Impurities (DNAses, RNases and Proteases):None
> Melting Point:88°C ± 1.5°C (1.5% gel)
> Moisture:≤ 10%
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Solubility:Clear colorless solution at 1g in 100 mL water
> Storage Condition:Ambient
> Sulfate:≤ 0.15%
Specifications
Top Vision Agarose Tablet | |
88 °C | |
200 Tablets (0.5 g each) |
100 to >30000 bp | |
TopVision | |
Gel Electrophoresis |
For Research Use Only. Not for use in diagnostic procedures.