Learn More
Thermo Scientific™ Flat PCR Cap Strips
Description
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Thermo Scientific Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
• Large end tabs for strip removal (12 strips only)
Autoclavable Resistance:Successfully Tested
> DMSO (100%) Resistance:Successfully Tested
> Ethanol Resistance:Successfully Tested
> Gamma Irradiation Resistance:Successfully Tested
> Hazardous:No
>Isopropanol Resistance:Successfully Tested
> Long Term Storage:Successfully Tested
> PCR (including waterbath):Successfully Tested
> Peelable:Successfully Tested
> Pierceable:Not Recommended
> qPCR:Not Recommended
> Resealable:Successfully Tested
> Sealing Temperature Range:-20°C to +120°C
> Storage Condition:Ambient
> UV Irradiation Resistance:Successfully Tested
Certifications
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
| Certifications/Compliance | Certified free from human DNA, RNases, and DNases |
| Quantity | 300 |