Learn More
Thermo Scientific™ Forma™ Humidity Pan
Material: Copper; For Use With: Forma Steri-cycle CO2 Incubator, Forma Series 310 Direct heat CO2 Incubator, Forma series II Water jacketed CO2 Incubator
Brand: Thermo Scientific™ 237020
This item is not returnable.
View return policy
Description
Copper humidity pan for use with Forma™ incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
For use with:
- Forma™ Steri-cycle CO2 incubator
- Forma Series 310 Direct heat CO2 incubator
- Forma Series II Water-jacketed CO2 incubator
Specifications
Forma Steri-cycle CO2 incubator, Forma Series 310 Direct heat CO2 incubator, Forma series II Water jacketed CO2 incubator | |
Copper | |
Humidity Pan |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
20Minutes
7Seconds
0
Product Content Correction
Product Title
Thermo Scientific™ Forma™ Humidity Pan > Copper Humidity Pan
Spot an opportunity for improvement?Share a Content Correction