Learn More
Thermo Scientific™ GeneJET FFPE DNA Purification Kit
Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue.
Brand: Thermo Scientific™ K0882
559.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific GeneJET FFPE DNA Purification Kit is designed for fast and convenient purification of DNA from various amounts of archived FFPE tissue. Up to 8 sections (10 μm thickness) of FFPE sample from various tissues can be used for genomic DNA extraction without overnight incubation. Elimination of toxic reagents commonly used for deparaffinization (i.e., xylene) allows a safe DNA purification procedure with high DNA yields and excellent quality. Each preparation recovers up to 8 μg of genomic DNA that can be eluted in volumes from 20 μL allowing for concentrated samples. Due to high decrosslinking efficiency high quality purified DNA shows reliable performance in sensitive downstream applications such as qPCR, PCR (amplicons up to 1 kb) or NGS library preparation.
Highlights
• High quality DNA—suitable for PCR and qPCR
• Fast—procedure can be completed within 2 hours
• Safe—non toxic solvents
• Efficient—yields up to 8 μg of purified DNA
Applications
• PCR
• qPCR
• NGS library preparation
Includes
• Proteinase K Solution
• RNase A Solution
• Digestion Buffer
• Binding Buffer
• Wash Buffer I (conc.)
• Wash Buffer II (conc.)
• Elution Buffer
• GeneJET DNA Purification Columns & Collection Tubes
• Collection Tubes
• Detailed Protocol
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifications
Spin Column | |
Spin Column | |
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), PCR | |
250 Preps | |
FFPE DNA Purification Kit | |
Genomic DNA |
Spin Column | |
FFPE, Fixed Samples | |
Not High-throughput Compatible (Manual) | |
GeneJET | |
250 Preps | |
<20 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.