missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reagents for GeneJET™ Plasmid Miniprep Kit

Product Code. 10202510 Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.7 mL
Product Type:
Ribonuclease A
Unit Size:
0.7mL
This item is not returnable. View return policy

Product Code. 10202510

Brand: Thermo Scientific™ R1253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Components of the GeneJET Plasmid Miniprep Kit (K0502/K0503) that may be purchased separately.

Thermo Scientific™ GeneJET™ Neutralization Solution and Lysis Solution are components of the GeneJET Plasmid Miniprep Kit (K0502/K0503) and may be purchased separately.

The kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20μg of high copy plasmid DNA per isolation procedure.

The Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Highlights
  • Efficient - high yields of up to 20 μg of high quality plasmid DNA
  • Fast - procedure takes less than 14 minutes
  • Convenient - no phenol/chloroform extraction or alcohol precipitation required
  • Pure - purified DNA is immediately ready to use
Applications
Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
  • FastDigest or conventional restriction digestion
  • Automated fluorescent and radioactive sequencing
  • PCR
  • In vitro transcription
  • Transformation
Principle
A bacterial culture is harvested and lysed. The lysate is then cleared by centrifugation and applied on the silica column to selectively bind DNA molecules at a high salt concentration. The adsorbed DNA is washed to remove contaminants, and the pure plasmid DNA is eluted in a small volume of elution buffer or water. The purified DNA is ready for immediate use in all molecular biology procedures, such as fast and conventional digestion with restriction enzymes, PCR, in vitro transcription, transformation, and automated sequencing.

Components
  • Resuspension Solution
  • Lysis Solution
  • Neutralization Solution
  • Wash Solution (concentrated)
  • RNase A Solution
  • Elution Buffer
  • Spin Columns
  • Collection Tubes
  • Detailed Protocol

Hazardous: No
Quality Control:

  • The kit is tested for the isolation of pUC19 DNA from E. coli according to the protocol.
  • The quality of the isolated DNA is evaluated by spectrophotometric assay, agarose gel electrophoresis, digestion with FastDigest restriction enzymes, and automated sequencing.
Storage Condition: Ambient,-20°C

Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:—FastDigest or conventional restriction digestion,—automated fluorescent and radioactive sequencing

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature,-20°C
For Use With (Application) DNA extraction
Product Type Ribonuclease A
Quantity 0.7 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.