Learn More
Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Cylindrical Longpath Cell Holder
Brand:  Thermo Scientific™ 336012-000
Product Code. 11873754
  • £250.00 / Each
Explore more special offers
Description
Description
For GENESYS 10 Spectrophotometers with single-cell cuvet holder.
  • Accommodates a cylindrical cell with a 10 to 100mm pathlength, 22 to 25mm dia. and Z dimension of 11mm Models with six-position cell holders can accommodate single-cell holders using accessory Single-cell Platform and Holder (14-385-476), available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cylindrical Cell Holder
Genesys™ 10 UV-vis spectrophotometer
100 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Cylindrical Longpath Cell Holder > Flow Cell, 18 micron L