Learn More
Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Test Tube Holder
Product Code. 11851872 Shop All Thermo Scientific Products
Click to view available options
Type:
Test Tube Holder
Unit Size:
Each
Product Code. 11851872
Brand: Thermo Scientific™ 336014000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
  • Accommodates one 25 dia. × 102mmH test tube or COD reagent vial
  • For GENESYS 10 spectrophotometers with six-position cell holder (holds three) or single-cell holder platform (holds one)
TRUSTED_SUSTAINABILITY
Specifications
Type Test Tube Holder
For Use With (Equipment) Genesys™ 10 UV-vis spectrophotometer
Holds One 25 dia. x 102 mm H test tube or COD reagent vial