Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Cylindrical Longpath Cell Holder
Brand: Thermo Scientific™ 4011-000
This item is not returnable.
View return policy
Description
Cylindrical Longpath Cell Holder. Snap-in” holder accepts one cylindrical longpath cell up to 50mm long and 22mm in diameter.
Specifications
Cylindrical Cell Holder | |
Genesys™ 20 spectrophotometer | |
Up to 50 mm |