Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Rectangular Longpath Cell Holder
Brand:  Thermo Scientific™ 4012-000
Product Code. 10490001
  • £194.00 / Each
Explore more special offers
Description
Description
Rectangular Longpath Cell Holder. Easy-to-install “snap-in” cell holder accommodates one rectangular longpath cell up to 50mm long and 12mm wide.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell Holder
Cell up to 50m L and 12mm W
Rectangular
Genesys™ 20 spectrophotometer
50 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Rectangular Longpath Cell Holder > Cell Holder, 50mm L