Learn More
Thermo Scientific™ Heracell™ External Gas Tank Guard
For Use With: 150i and 240i Incubators
Brand:  Thermo Scientific™ 50059043
Additional Details : Weight : 0.10000kg
Product Code. 17135224
  • £1110.03 / Each
In Stock
to see stock.
Add to basket
Description
Description
Accessory for Steri-Cycle i160 CO2 incubators
  • External gas guard automatic change-over to reserve tank
  • 120 V, 50/60 Hz
Specifications
Specifications
150i and 240i incubators
Accessory, External Gas Tank Guard
120 V
LED
120 V, 50/60 Hz
50/60 Hz