Learn More
Thermo Scientific™ Hypercarb™ Preparative Guard Cartridges
Use preparative guard cartridges to extend column lifetime
Brand:  Thermo Scientific™ 35005-019023
Code : A0
Additional Details : Weight : 0.10000kg
Product Code. 11593201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Suited to solve problem separations
  • Exceptional retention of very polar analytes
  • True orthoganality to C18
  • Separated structurally related substances
  • Excellent for high temperature applications
Specifications
Specifications
5 μm
10 mm
10 mm
Hypercarb