Learn More
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
Brand:  Thermo Scientific™ 28105-012106
388.85 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11528710
  • £390.95 / Pack of 4
Explore more special offers
Description
Description
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of the BDS C18 Solid Phase
  • Base deactivated with minimal residual silanol activity
  • Economical with exceptional reproducibility
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 μm
10 mm
2.1 mm
C18
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Hypersil™ BDS C18 Javelin Guard Columns > Particle Size: 5μm; 10L x 2.1mm I.D.