Learn More
Thermo Scientific™ Hypersil™ BDS C8 Reversed Phase HPLC Column, 3 μm, 3 mm x 50 mm
Column Format: Analytical Column; Carbon Load: 7%; Pore Size: 130 Å; Column Phase: Reversed Phase
Brand: Thermo Scientific™ 28203-053030
Code : 16
Additional Details : Weight : 0.10000kg
Description
- Excellent choice for acids, bases and neutral compounds
- Less retentive than C18 for faster analyses
- Robust columns with long column lifetimes
- Very reliable for routine assays
Order Info
Other column dimensions are available. Please call Customer Service for more information.
Specifications
Analytical Column | |
50 mm | |
3 μm | |
130 Å | |
60°C | |
C8 | |
7% | |
Yes |
3 mm | |
5800 psi (400 bar) | |
2 to 8 | |
170 m²/g | |
L7 | |
Spherical, Fully Porous, Base-Deactivated Silica | |
Reversed Phase | |
Reversed Phase |