Learn More
Thermo Scientific™ Labtips Pipette Tips
Produced from the highest quality virgin polypropylene
Brand:  Thermo Scientific™ 94300230
116.00 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 4.79000kg
 View more versions of this product
Product Code. 12336582
  • £218.00 / Pack of 1800
In Stock
to see stock.
Add to basket
Description
Description
  • Double injection molded for maximum sample delivery and best fit for a variety of pipetters
  • Available in 200 and 1000μL including bulk and racked formats
Specifications
Specifications
1000 μL
Blue
Rack
LabTip Pipette
Polypropylene
Non-sterile
Autoclavable
Universal Fit
LabTip