Learn More
Unity Lab Services™ Thermo Scientific™ Liquid Nitrogen Storage Equipment Validation Documention for Thermo Scientific™ CryoMed™ Controlled-Rate Freezers
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Liquid Nitrogen Storage Equipment Validation Documention for Thermo Scientific CryoMed™ Controlled-Rate Freezers
Brand: Unity Lab Services™ IOQDOCE89003505
Additional Details : Weight : 0.00010kg
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Thermo Scientific Liquid Nitrogen Storage Equipment including Thermo Scientific CryoMed™ Controlled-Rate Freezers | |
Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
IQ/OQ Documentation |