Learn More
Thermo Scientific™ Low Profile Tubes (white) and Ultra Clear Caps, strips of 8
Low profile strip tubes for PCR and qPCR applications.
Brand: Thermo Scientific™ AB1771
Description
The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.
The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.
Highlights
- Suitable for 0.2 mL thermal cycler blocks
- 150 μL maximum tube capacity when capped
- Low profile to reduce dead space and increase PCR efficiency
- Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
- PCR
- qPCR/Real-Time PCR
Hazardous: No
Storage Condition: Ambient
PCR, qPCR and Real-Time PCR.
Certifications
Certified free from human DNA, RNases, and DNases.

Specifications
Certified DNA/RNase Free | |
0.1 mL | |
250 strips | |
Non-sterile | |
Low profile | |
Ultra-clear Caps | |
Low Profile Tube and Ultra Clear Cap |
White, Ultra Clear | |
0.15 mL | |
Thermal Cyclers or Real-Time PCR Systems | |
Polypropylene | |
Flat Cap | |
250 Strip Tubes and Caps |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.