missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima H Minus Reverse Transcriptase (200 U/μL)

Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT.

Brand:  Thermo Scientific™ EP0752

160.00 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 13233159

  • £240.00 / 10000 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• Maxima H Minus Reverse Transcriptase, 1 x 10,000 units (200 U/μL)
• 5X RT Buffer

Store at –20°C.

High
Reverse Transcription
10,000 Units
Reduced
200 U/μL
Separate components
Up to 20 kb
Stand-alone Enzyme
15 to 30 min.
50°C to 55°C
Maxima H Minus
Dry Ice
First-Strand cDNA
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2812537 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0752
Lot Number2847437 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0752
Lot Number2876054 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0752
Lot Number2912784 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberEP0752
Lot Number2912772 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberEP0752
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Maxima H Minus Reverse Transcriptase (200 U/μL) > 10,000 Units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.