Learn More
Thermo Scientific™ Maxima H Minus Reverse Transcriptase (200 U/μL)
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT.
£71.25 - £750.00
Specifica
Format | Stand-alone Enzyme |
---|---|
GC-Rich PCR Performance | High |
Reaction Speed | 15 to 30 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 50°C to 55°C |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
13223159
|
Thermo Scientific™
EP0751 |
2,000 Units |
£71.25
2000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13233159
|
Thermo Scientific™
EP0752 |
10,000 Units |
£240.00
10000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13243159
|
Thermo Scientific™
EP0753 |
4 x 10,000 Units |
£750.00
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT. The engineered enzyme features dramatically improved thermostability, 50X higher processivity, robustness, and increased synthesis rate compared to wild type M-MuLV RT.
The eliminated RNase H activity enables the enzyme to produce very long RNA transcripts up to 20 kb. Due to its high thermostability, the enzyme maintains full activity during the entire reverse transcription reaction and generates high yields of cDNA. The reaction temperature can be increased to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. The extremely high processivity of Maxima H Minus enzyme results in increased resistance to common reaction inhibitors, such as guanidine, formamide, and ethanol.
Features of Maxima H Minus Reverse Transcriptase include:
• Thermostable—90% active after incubation at 50°C for 60 minutes in a reaction mixture
• Active up to 65°C
• High yields of full-length cDNA up to 20 kb
• High sensitivity—reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
• Efficient—complete cDNA synthesis in 15 to 30 minutes
• Increased resistance to common reaction inhibitors
• Incorporates modified nucleotides
Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Reverse transcription at elevated temperatures to reduce effects of secondary structure
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• Analysis of RNA by primer extension
Related products
• Maxima H Minus Reverse Transcriptase (Cat. No. EP0752)
• Maxima H Minus First Strand cDNA Synthesis Kit (Cat. No. K1652)
• Maxima H Minus First Strand cDNA Synthesis Kit, with dsDNase (Cat. No. K1681)
• Maxima H Minus cDNA Synthesis Master Mix (Cat. No. M1661)
• Maxima H Minus cDNA Synthesis Master Mix, with dsDNase (Cat. No. M1681)
Order Info
Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.
Specifica
Stand-alone Enzyme | |
15 to 30 min. | |
50°C to 55°C | |
Reduced | |
200 U/μL | |
Separate components | |
Up to 20 kb |
High | |
Reverse Transcription | |
Maxima H Minus | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.