missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Microplate Carrier for H-6000A Buckets
Carriers for H-6000A, HBB-6, HLR-6 swinging bucket rotors
Brand: Thermo Scientific™ 11267
This item is not returnable.
View return policy
Description
- Holds 6 x 3 standard or 6 x 1 deepwell microplates
- Maximum tube dimensions: 60 x 127mm
- Maximum Speed: 5000rpm
- Maximum RCF: 5186xG
Specifications
Microplate Carrier | |
H-6000A Bucket | |
6 x 3 Standard or 6 x 1 Deepwell Microplates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction