Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles Product Code.: 10050561
In Stock
to see stock.
Add to basket
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
GREENER_CHOICE
Product Code. 10050561
28-400
Click to view available options
Neck Finish:
24-400
28-400
Product Code. 10050561
Brand: Thermo Scientific™ 7124010001
In Stock
to see stock.
Add to basket
Add to basket
Through-the-top tubing with closure and stem molded in one piece
  • Closure and stem molded in one piece for leakproof service
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Leakproof†
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Neck Finish 28-400
For Use With (Equipment) Wash bottle
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number7450424010 Certificate TypeCertificate of Compliance Date25/02/2025 Catalog Number712401-0001
Lot Number7446429010 Certificate TypeCertificate of Compliance Date13/01/2025 Catalog Number712401-0001
Lot Number7444990010 Certificate TypeCertificate of Compliance Date12/01/2025 Catalog Number712401-0001
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles >