Learn More
Thermo Scientific™ Nalgene™ System 100™ Cryogenic Vials
Externally threaded polypropylene self-standing vials
£423.36 - £457.70
Specifications
Material Polypropylene
Sterility Sterile
Thread Style External
Quantity 500/Cs.
View More Specs
Alert:
Cryogenic vials are intended for placement only in the vapor phase of liquid nitrogen, and should not be used for storage in the liquid phase of liquid nitrogen. Immersion of the vials in the liquid phase could result in penetration of the liquefied gas into the vial, resulting in rapid vaporization of the liquid upon removal and possible violent explosion or leakage from the vial/closure perimeter.
To prevent cryogenic vials from exploding, never overfill liquid nitrogen storage units. Always examine vials before use to ensure no visible defects around the closure rims. Always use full faceshields, heavy safety gloves and laboratory protective apparel when removing vials from cryogenic storage. Always permit vials to warm slowly in a biological safety cabinet. Never reuse cryogenic vials.
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
13417659
View Documents
Thermo Scientific™
AY509X32
1 mL
£457.70
Pack of 500
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
12784716
View Documents
Thermo Scientific™
AY509X33
1.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Sterile
  • For use with System 100 CryoBoxes™ and 25- and 81-ampule boxes with 2.0mL ampule capacities
Specifications
Specifications
Polypropylene
Sterile
External
500/Cs.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ System 100™ Cryogenic Vials