Learn More
Thermo Scientific™ Nunc™ Caps for TufRol™ Roller Bottles
Click to view available options
Closure Type:
Non-vented caps
Vented
Description
Vented and non-vented
- Molded of high density polyethylene (HDPE)
- Designed to prevent misthreading and to provide a secure seal
- Vented cap has an integral hydrophobic 0.2μm microporous membrane filter vent, allowing consistent gas exchange when cap is in the closed position
Specifications
Specifications
No. per Case | 500 Cs. |
Sterility | Sterile |
For Use With (Equipment) | TufRol roller bottle |
Closure Type | Vented |
Product Content Correction
Product Title
Thermo Scientific™ Nunc™ Caps for TufRol™ Roller Bottles >
Spot an opportunity for improvement?Share a Content Correction