Learn More
Fisherbrand™ accumet™ Chloride Combination Electrode - Mercury-Free
Typical applications include water/wastewater, soil, dairy, meats, tomato/vegetable products.
Brand:  Fisherbrand™ 93X218742
619.49 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 11500154
  • £772.00 / Each
Estimated Shipment: 11-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Typical applications include water/wastewater, soil, dairy, meats, tomato/vegetable products.
Method ASTM/AOAC approved
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chloride
BNC
Plastic
Chloride Combination Electrode
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ accumet™ Chloride Combination Electrode - Mercury-Free > Chloride; Combination