missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters

Use these calibration standards to check your colorimeter instrument operations before measuring COD chemistries, for example in environmental and wastewater monitoring.

Brand:  Thermo Scientific™ CODS01

 View more versions of this product

Product Code. 10316522

  • £86.45 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.

  • For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ2040
  • Two Calibration Standards: 1000 ppm and 10000ppm, both in 475 mL bottles

Order Info

Reagents sold separately.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1000ppm
AQUAfast Colorimeter
Environmental and Wastewater Monitoring
475 mL
EPA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers