missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ ADJ1 Adjustable System Accessories and Replacement Parts

Extremely compact at only 16cm wide

Brand:  Thermo Scientific™ ADJ1-40R

 View more versions of this product

Product Code. 11510305

  • £52.95 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The height of the upper buffer chamber (UBC) may be adjusted by sliding the UBC up or down two support poles allowing gels to vary in height from 14 cm to 40 cm.
  • The notched or offset glass plate of the gel sandwich presses against a silicone gasket in the UBC for a leakproof seal.
  • Plates are clamped to the UBC with either binder clamps or heavy duty pony clamps.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Owl ADJ1 Adjustable Electrophoresis System
Glass
0.48 cm
15.7 in.
6.2 in.
16 cm
Notched Glass Plate
0.18 in.
15.7 x 6.2 in. (40 x 16 cm)
40 cm
6.2 in.
16 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers