Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Influenza A & B
For the detection and identification of Influenza virus strains A and B in clinical specimens or cell cultures
Brand: Thermo Scientific™ K610511-2
601.33 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Test utilizes species-specific monoclonal antibodies to detect epitopes of Influenza virus glycoproteins and fusion proteins specific to either Influenza virus type A or Influenza virus type B
Specifications
50 Tests |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ IMAGEN™ Influenza A & B > 50 tests
Spot an opportunity for improvement?Share a Content Correction