Learn More
Thermo Scientific™ Phusion U Multiplex PCR Master Mixes
Thermo Scientific Phusion U Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube.
£240.00 - £940.00
Specifications
Polymerase | Phusion U Hot Start DNA Polymerase |
---|---|
Reaction Speed | Fast |
Product Type | Multiplex PCR Master Mix |
Shipping Condition | Dry Ice |
Concentration | 2X |
Product Code | Brand | Color | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Reactions | Price | Quantity & Availability | ||||
15371722
|
Thermo Scientific™
F564S |
Green | 100 Reactions |
£240.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15351722
|
Thermo Scientific™
F562L |
Colorless | 500 Reactions |
£934.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15341722
|
Thermo Scientific™
F562S |
Colorless | 100 Reactions |
£242.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15381722
|
Thermo Scientific™
F564L |
Green | 500 Reactions |
£940.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific Phusion U Green Multiplex PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.
The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Green format further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.
Highlights
- Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
- Excellent results with templates of suboptimal purity due to inhibitor tolerance
- Fast cycling and direct loading of PCR products on gels
Applications
- Genotyping
- Pathogen detection
- Food testing
- Analysis of genetically modified organisms
- Amplification of microsatellites
The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mixes can even be used with unpurified samples such as blood or serum.
Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.
Using Phusion DNA Polymerases
Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).
Specifications
Phusion U Hot Start DNA Polymerase | |
Multiplex PCR Master Mix | |
2X | |
Built-In Hot Start | |
SuperMix or Master Mix |
Fast | |
Dry Ice | |
25X | |
Blunt | |
2.5 kb or less |
For Research Use Only. Not for use in diagnostic procedures.