missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Anti-HA Agarose

Product Code. 11543060 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mL
5 mL
missing translation for 'unitSize'
10mL
Each
This item is not returnable. View return policy

Product Code. 11543060

missing translation for 'mfr': Thermo Scientific™ 26182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HA Epitope Tag Monoclonal antibody 26181 and 26182 from Thermo Scientific™ specifically detects HA Epitope Tag in Tag samples, and it is validated for Immunoprecipitation

An HA tag is frequently incorporated into recombinant proteins for a variety of purposes. An anti-HA antibody can then be used to detect the protein when doing studies with transfected cells.
Host Species: Mouse
Cross Reactivity: Tag
Isotype: IgG1
Clone: 2-2.2.14
Immunogen: HA peptide YPYDVPDYA derivitized to ovalbumin
Formulation: 10mL Purified Antibody in PBS, with 0.05% sodium azide; pH7.2
Synonyms: human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
26182 is supplied as 10mL of antibody on crosslinked 4% beaded agarose supplied as a 50% slurry in 0.1M phosphate, 0.15M NaCl, pH 7.2 with 0.05% sodium azide. Specific for the HA peptide YPYDVPDYA. The anti-HA agarose has been used successfully in immunoprecipitation applications. The anti-HA agarose has 3.5mg mouse anti-HA IgG1 monoclonal antibody conjugated permL of settled agarose resin. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Agarose
Column Type Agarose Resin, Affinity
Format Bottle
Conjugate HA-epitope tag (YPYDVPDYA)
Purification Target Anti-HA Antibody
Content And Storage 5 mL of settle resin, provided as 10 mL of 50% slurry. Store at 2–8°C.
Quantity 5 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.