Learn More
Thermo Scientific™ HA Synthetic Peptide
HA Synthetic peptide 26184 from Thermo Scientific™ specifically detects HA in samples, and it is validated for Control
Brand: Thermo Scientific™ 26184
257.75 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : CAS Number : 92000-76-5 Weight : 0.45360kg
Description
HA Synthetic peptide 26184 from Thermo Scientific™ specifically detects HA in samples, and it is validated for Control
The HA peptide, NH2-YPYDVPDYA-COOH, is a synthetic peptide used to competitively elute fusion proteins containing the influenza hemagglutinin (HA) tag bound to immobilized anti-HA affinity resins.Formulation: 5 mg purified Peptide (1mg/mL) no preservative;
26184 detects HA in samples. 26184 has been successfully used in Control procedures.
Specifications
Recombinant | |
Store at 4°C. | |
Immunoprecipitation | |
5 mg |
Safety and Handling
- Isopropanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : isopropanol
For Research Use Only. Not for use in diagnostic procedures.