missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ B-PER™ Complete Bacterial Protein Extraction Reagent

Product Code. p-133431 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15255743

Brand: Thermo Scientific™ 89821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Reagent packages are available in Tris and phosphate buffer formulations; Kits contain Tris-based B-PER Reagent plus lysozyme and DNase I enzymes.

Efficient, gentle lysis and extraction of E.coli and other bacterial cells

  • An easy-to-use cell lysis reagent that effectively disrupts cells and solubilizes native or recombinant proteins without denaturation
  • All B-PER Reagents are compatible with downstream applications such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay)
  • This all-inclusive formulation combines the lysis reagent with lysozyme, and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria
  • Depending on the particular application protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.

Features of the B-PER Bacterial Protein Extraction Reagent:

  • Detergent in Tris buffer with Lysozyme and Universal Nuclease
  • Improved cell lysis and DNA digestion, thereby releasing soluble proteins and reducing viscosity to increase yields
  • Recovery of large molecular weight proteins (high MW proteins)
  • Cost-effective equivalent to Merck Millipore Bugbuster™ Master Mix
  • More effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications
TRUSTED_SUSTAINABILITY

Specifications

Formulation Proprietary detergent in 20mM Tris buffer, pH 7.5, containing lysozyme and universal nuclease
Quantity 250 mL
Product Type Bacterial Protein Extraction Reagent
Volume (Metric) 250 mL
Content And Storage Store at 4°C.
Product Line B-PER

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.