Learn More
Thermo Scientific™ Pierce™ BSA Protein Digest, MS grade
Calibrate and maintain quality control for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples using the carefully validated Thermo Scientific™ Pierce™ BSA Protein Digest Standard, LC-MS Grade.
Brand: Thermo Scientific™ 88341
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The Thermo Scientific Pierce BSA Protein Digest Standard, LC-MS Grade, is a validated protein digest optimized for use as a quality control sample for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples.
- Positive control sample - BSA digest optimized and validated as a quality control standard for mass spectrometry applications
- Excellent sequence coverage - greater than 85% sequence coverage
- Validated peptide quality - digestion procedure optimized for minimal missed cleavages and over-alkylation
- Rigorously tested - high-quality, consistent BSA protein digest documented via lot-specific certificates of analysis
- Stable - provided in a stable lyophilized format
Specifications
Mass Spectrometer | |
Pierce™ | |
Peptides | |
Mass Spectrometry | |
Solid |
Protein Digest | |
Store in freezer (-5 to -30°C). | |
Peptides | |
Calibration |
For Research Use Only. Not for use in diagnostic procedures.