missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt

Firefly luciferase substrate for in vitro and in vivo applications

Brand:  Thermo Scientific™ 88293

 View more versions of this product

Product Code. 15215733

  • £112.00 / 100mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce D-Luciferin is formulated at greater than 99% purity as both monosodium and monopotassium salts to provide convenience and high performance at a low cost in reporter assays involving firefly luciferase.

Luciferin is the substrate for bioluminescence reactions with firefly luciferase enzyme. Lyophilized potassium and sodium salts of firefly luciferin are stable and easily soluble, and both formulations are compatible with a wide variety of luciferase-based in vivo imaging and in vitro bioluminescent assays. (The decision to use one or the other form is a matter of researcher preference.) Applications include ATP assays, gene reporter assays, high-throughput sequencing and various contamination assays throughout the biotechnology field. For in vivo imaging, the injection of luciferin in model systems enables real-time, non-invasive monitoring of disease progression and/or drug efficacy through bioluminescence.

Highlights:

  • High purity, typically greater than 99.7%, as verified by HPLC and FTIR
  • Great performance with in vitro applications
  • Suitable for in vivo imaging
Applications:
  • Gene reporter assays
  • ATP assays
  • High throughput sequencing
  • Contamination assays
  • Bioluminescent in vivo imaging
  • Drug screening
  • Multiplexed assays to study off-target effects
  • Signal transduction pathway analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at or below -20°C.
ATP Assay, Gene Reporter Assay
Pierce™
Chemical Substrate
Bioluminescence
D-Luciferin, Monopotassium Salt
100 mg
Luciferase Substrate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.